2-methyl-4-(trifluoromethoxy)phenol 97%

Warning

H33957-MD H33957-03
AAH33957-03EA 133.2 USD
AAH33957-03 AAH33957-MD
2-methyl-4-(trifluoromethoxy)phenol 97%
2-methyl-4-(trifluoromethoxy)phenol
Formula: C₈H₇F₃O₂
MW: 192.137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972927
CAS Number: 129676-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR