2-Fluoro-4-nitrobenzaldehyde 97%
|
Warning
|
H33937-03
H33937-MD
AAH33937-MDEA
33.08
USD
AAH33937-MD
AAH33937-03
2-Fluoro-4-nitrobenzaldehyde 97%
2-Fluoro-4-nitrobenzaldehyde
Formula:
C₇H₄FNO₃ MW: 169.10 g/mol Melting Pt: 98…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00968941 CAS Number: 157701-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...