4-Chloro-2-fluoro-3-methoxybenzylamine 97%
|
Danger
|
H33936-03
H33936-MD
AAH33936-03EA
136.53
USD
AAH33936-03
AAH33936-MD
4-Chloro-2-fluoro-3-methoxybenzylamine 97%
4-Chloro-2-fluoro-3-methoxybenzylamine
Formula:
C₈H₉ClFNO Storage Temperature: Ambient |
MDL Number:
MFCD19687209 CAS Number: 1323966-34-2 UN: 3259 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...