4-Hydroxy-3-iodobenzotrifluoride 97%
|
Danger
|
Synonyms:
2-Iodo-4-(trifluoromethyl)phenol
H33928-MD
H33928-03
AAH33928-MDEA
76.01
USD
AAH33928-MD
AAH33928-03
4-Hydroxy-3-iodobenzotrifluoride 97%
4-Hydroxy-3-iodobenzotrifluoride
Formula:
C₇H₄F₃IO MW: 288.01 g/mol Boiling Pt: 105 °C/44 mm Storage Temperature: Ambient |
MDL Number:
MFCD12922960 CAS Number: 463976-21-8 UN: 3264 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...