2'-Methyl-4'-(trifluoromethoxy)acetophenone 97%
|
Warning
|
Synonyms:
2-Methyl-4-(trifluoromethoxy)acetophenone
H33870-03
H33870-MD
AAH33870-MDEA
25.95
USD
AAH33870-MD
AAH33870-03
2'-Methyl-4'-(trifluoromethoxy)acetophenone 97%
2'-Methyl-4'-(trifluoromethoxy)acetophenone
Formula:
C₁₀H₉F₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD22201061 CAS Number: 1373920-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...