3-[2-Methyl-4-(trifluoromethoxy)phenyl]propionic acid 97%
|
Warning
|
Synonyms:
3-[2-Methyl-4-(trifluoromethoxy)phenyl]propanoic acid
H33829-03
H33829-MD
AAH33829-03EA
133.2
USD
AAH33829-03
AAH33829-MD
3-[2-Methyl-4-(trifluoromethoxy)phenyl]propionic acid 97%
3-[2-Methyl-4-(trifluoromethoxy)phenyl]propionic acid
Formula:
C₁₁H₁₁F₃O₃ MW: 248.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22201060 CAS Number: 1373921-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...