3-(Pentafluorothio)benzenesulfonyl chloride 97%

Danger

Synonyms: 3-(Pentafluorosulfur)benzenesulfonyl chloride3-(pentafluoro-lambda-sulfanyl)benzene-1-sulfonyl chloride

H33799-03 H33799-MD
AAH33799-03EA 198.69 USD
AAH33799-03 AAH33799-MD
3-(Pentafluorothio)benzenesulfonyl chloride 97%
3-(Pentafluorothio)benzenesulfonyl chloride
Formula: C₆H₄ClF₅O₂S₂
MW: 302.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652436
CAS Number: 1211520-62-5
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR