3-Methoxy-2-(trifluoromethyl)benzyl alcohol 97%
|
Warning
|
H33709-MD
H33709-03
AAH33709-03EA
178.71
USD
AAH33709-03
AAH33709-MD
3-Methoxy-2-(trifluoromethyl)benzyl alcohol 97%
3-Methoxy-2-(trifluoromethyl)benzyl alcohol
Formula:
C₉H₉F₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD18398589 CAS Number: 1261750-58-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...