4-Chloro-3-ethoxy-2-fluorobenzylamine 97%

Danger

H33498-03 H33498-MD
AAH33498-MDEA 92.59 USD
AAH33498-MD AAH33498-03
4-Chloro-3-ethoxy-2-fluorobenzylamine 97%
4-Chloro-3-ethoxy-2-fluorobenzylamine
Formula: C₉H₁₁ClFNO
Storage Temperature: Ambient
MDL Number: MFCD19687221
CAS Number: 1323966-28-4
UN: 3259
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR