2-Fluoro-5-methyl-4-(trifluoromethyl)benzylamine 97%

Danger

H33411-03 H33411-MD
AAH33411-03EA 236.43 USD
AAH33411-03 AAH33411-MD
2-Fluoro-5-methyl-4-(trifluoromethyl)benzylamine 97%
2-Fluoro-5-methyl-4-(trifluoromethyl)benzylamine
Formula: C₉H₉F₄N
Storage Temperature: Ambient
MDL Number: FCD19687196
CAS Number: 1323966-36-4
UN: 3259
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR