2-Methyl-4-(trifluoromethoxy)phenylacetonitrile 97%
|
Warning
|
Synonyms:
2-[2-methyl-4-(trifluoromethoxy)phenyl]acetonitrile
H33235-03
H33235-MD
AAH33235-MDEA
46.2
USD
AAH33235-MD
AAH33235-03
2-Methyl-4-(trifluoromethoxy)phenylacetonitrile 97%
2-Methyl-4-(trifluoromethoxy)phenylacetonitrile
Formula:
C₁₀H₈F₃NO MW: 215.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07368719 CAS Number: 886763-09-3 UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...