3-Methoxy-4-(trifluoromethyl)benzoyl chloride 97%
|
Danger
|
Synonyms:
Benzoyl chloride, 3-methoxy-4-(trifluoromethyl)-
H33164-03
H33164-06
AAH33164-06EA
496.17
USD
AAH33164-06
AAH33164-03
3-Methoxy-4-(trifluoromethyl)benzoyl chloride 97%
3-Methoxy-4-(trifluoromethyl)benzoyl chloride
Formula:
C₉H₆ClF₃O₂ MW: 238.59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18398587 CAS Number: 1261571-92-9 UN: 3261 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...