(4-Methoxy-6-methyl-2-pyrimidinyl)thiourea ≥97%

Warning

Synonyms: N-(4-Methoxy-6-methylpyrimidin-2-yl)thiourea(4-methoxy-6-methylpyrimidin-2-yl)thiourea1-(4-methoxy-6-methylpyrimidin-2-yl)thiourea

H33090-03 H33090-MD H33090-06
AAH33090-03EA 184.15 USD
AAH33090-03 AAH33090-06 AAH33090-MD
(4-Methoxy-6-methyl-2-pyrimidinyl)thiourea ≥97%
(4-Methoxy-6-methyl-2-pyrimidinyl)thiourea
Formula: C₇H₁₀N₄OS
MW: 198.15 g/mol
Melting Pt: 213…217 °C
Storage Temperature: Ambient
MDL Number: MFCD03291910
CAS Number: 93744-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR