2-Methyl-4-(trifluoromethoxy)cinnamic acid 97%

Warning

H33079-MD H33079-03
AAH33079-MDEA 41.22 USD
AAH33079-MD AAH33079-03
2-Methyl-4-(trifluoromethoxy)cinnamic acid 97%
2-Methyl-4-(trifluoromethoxy)cinnamic acid
Formula: C₁₁H₉F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD18399792
CAS Number: 1588508-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR