Cerium(IV) oxide, doped with 18% zirconium 13% in water, nanoparticle dispersion <5.0 nm APS Number Weighted, high pH

47259-22 47259-36
AA47259-22EA 190.92 USD
AA47259-22 AA47259-36
Cerium(IV) oxide, doped with 18% zirconium 13% in water, nanoparticle dispersion <5.0 nm APS Number Weighted, high pH
Cerium(IV) oxide, doped with 18% zirconium
Storage Temperature: Ambient
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR