Diaion® PA412 (Cl) 1.1 meq/ml on Poly(styrene-divinylbenzene), 6% cross-linked, strongly basic porous type ion-exchange resin II
46984-A1
AA46984-A1EA 81.03 USD
AA46984-A1
Diaion® PA412 (Cl) 1.1 meq/ml on Poly(styrene-divinylbenzene), 6% cross-linked, strongly basic porous type ion-exchange resin II
Diaion® PA412 (Cl)
Pure water, nucleic acids and amino acids