Diaion® UBA100 (Cl) 1.3 meq/ml on Poly(styrene-divinylbenzene), cross-linked, strongly basic gel type ion-exchange resin
46938-A1
AA46938-A1EA 117.66 USD
AA46938-A1
Diaion® UBA100 (Cl) 1.3 meq/ml on Poly(styrene-divinylbenzene), cross-linked, strongly basic gel type ion-exchange resin
Diaion® UBA100 (Cl)
Power industry condensate demineralization, metal recovery, catalysis