Diaion® UBK10 (Na) 2.2 meq/ml on Poly(styrene-divinylbenzene), 10% cross-linked, strongly basic gel type ion-exchange resin
46922-A1
AA46922-A1EA 96.45 USD
AA46922-A1
Diaion® UBK10 (Na) 2.2 meq/ml on Poly(styrene-divinylbenzene), 10% cross-linked, strongly basic gel type ion-exchange resin
Diaion® UBK10 (Na)
Softening, metal recovery, amino acid separation, sugar refining