Sepabeads® SP207 synthetic adsorbent resin brominated PS-DVB P.R. 110 angstroms, highly porous type
Supplier: Thermo Scientific Chemicals
046848-A1
AA46848-A1EA
221.55
USD
AA46848-A1
Sepabeads® SP207 synthetic adsorbent resin brominated PS-DVB P.R. 110 angstroms, highly porous type
Sepabeads® SP207
Applications: Bromination of the aromatic ring provides increased hydrophobicity, and consequently, increased selectivity for hydrophobic molecules versus conventional styrene/DVB synthetic adsorbents like Diaion® HP20 resin.
Density:
1.19 Storage Temperature: Ambient |
CAS Number:
87924-01-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...