Diaion® WK11(H), ion exchange resin, weakly acidic porous type, 2.9 meq/ml on polymethacrylate
46738-A1
AA46738-A1EA 114.79 USD
AA46738-A1
Diaion® WK11(H), ion exchange resin, weakly acidic porous type, 2.9 meq/ml on polymethacrylate
Pharmaceuticals, dealkalination, metal recovery, antibiotics, sucrose purifications