Diaion® UBK555 (Ca) 2.0 meq/ml on Poly(styrene-divinylbenzene), 8% cross-linked, strongly basic gel type ion exchange fractionation resin
46731-A1
AA46731-A1EA 89.31 USD
AA46731-A1
Diaion® UBK555 (Ca) 2.0 meq/ml on Poly(styrene-divinylbenzene), 8% cross-linked, strongly basic gel type ion exchange fractionation resin
Diaion® UBK555 (Ca)
Chromatographic sugar fractionation , fructose/glucose separations, sugar alcohol separations