Diaion® UBK12 (Na) ion exchange resin 2.3 meq/ml on poly(styrene-divinylbenzene), 12% cross-linked, strongly acidic gel type
46671-A1
AA46671-A1EA 94.57 USD
AA46671-A1
Diaion® UBK12 (Na) ion exchange resin 2.3 meq/ml on poly(styrene-divinylbenzene), 12% cross-linked, strongly acidic gel type
Diaion® UBK12 (Na)
Softening, metal recovery, amino acid separation, sugar refining