Sepabeads® SP710 synthetic adsorbent resin poly(styrene-divinylbenzene) P.R. 90 angstroms, highly porous type
Supplier: Thermo Scientific Chemicals
THERMO FISHER SCIENTIFIC
AA46555-A1EA 226.99 USD
AA46555-A1
Sepabeads® SP710 synthetic adsorbent resin poly(styrene-divinylbenzene) P.R. 90 angstroms, highly porous type
Sepabeads® SP710
  • 250 µm Beads
Synthetic adsorbents for juice debittering and decolorizing. Also highest naringin capacity