Diaion® PA408 (Cl) 0.9 meq/ml on Poly(styrene-divinylbenzene), 4% cross-linked, strongly basic porous type ion-exchange resin II
46474-A1
AA46474-A1EA 69.37 USD
AA46474-A1
Diaion® PA408 (Cl) 0.9 meq/ml on Poly(styrene-divinylbenzene), 4% cross-linked, strongly basic porous type ion-exchange resin II
Diaion® PA408 (Cl)
Sugar refining/decolorizing, particularly beet sugar