Sepabeads® SP70 synthetic adsorbent resin poly(styrene-divinylbenzene) P.R. 70 angstroms, highly porous type
Supplier: Thermo Scientific Chemicals
THERMO FISHER SCIENTIFIC
AA46410-A1EA 190.99 USD
AA46410-A1
Sepabeads® SP70 synthetic adsorbent resin poly(styrene-divinylbenzene) P.R. 70 angstroms, highly porous type
Sepabeads® SP70
  • 250 µm Beads
Synthetic adsorbents for juice debittering and decolorizing