Phenylmagnesium bromide 3 M in ether

Danger


Packaged under argon in resealable ChemSeal™ bottles.

042859-AE
AA42859-AEEA 94.97 USD
AA42859-AE
Phenylmagnesium bromide 3 M in ether
Phenylmagnesium bromide
Formula: C₆H₅BrMg
Density: 1.14
Storage Temperature: Ambient
MDL Number: MFCD00000038
CAS Number: 100-58-3
UN: 3399
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR