Molecular sieve, tantalum oxide based (Ta-TMSI), mesoporous, powder 22 Angstrom

Warning

Synonyms: Ti-TMS1

42612-MD 42612-06 42612-03
AA42612-06EA 398.73 USD
AA42612-06 AA42612-MD AA42612-03
Molecular sieve, tantalum oxide based (Ta-TMSI), mesoporous, powder 22 Angstrom
Molecular sieve, tantalum oxide based (Ta-TMSI)

Application : Catalysis, gas adsorption, optoelectronic materials

Storage Temperature: Ambient
MDL Number: MFCD00131613
EINECS: 215-238-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR