Glassy carbon, Powder 1000-2000 µm Type2

Warning

Synonyms: Vitreous carbon

42519-18 42519-30
AA42519-30EA 715.18 USD
AA42519-30 AA42519-18
Glassy carbon, Powder 1000-2000 µm Type2
Glassy carbon
Formula: C
MW: 12.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR