Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glassy Carbon Tapered Crucibles for Crystal Growth, Alfa Aesar

Supplier: Alfa Aesar
40952-KT 40953-KT 40959-KT 40954-KT 40957-KT 40958-KT 40956-KT 40955-KT
Glassy carbon offers high purity, corrosion resistance, thermal stability, and a structure impermeable to both gases and liquids

  • Resistance to all wet decomposition agents
  • No memory effects (uncontrolled adsorption and release of foreign elements)
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Good resistance to thermal shock allows rapid heating and cooling times
  • Resistive to abrasive wear
  • Good electrical conductivity

These crucibles are designed for high-temperature differential thermal/thermal gravimetric analyses and ultra-high purity materials technology such as crystal growing.

Alta Aesar's glassy carbon products are subjected to a high temperature heat treatment which imparts special material properties, including significantly improved corrosion resistance and strength.




 

ORDER

Expand All / Collapse All
Capacity Height Top Diameter Supplier No. VWR Catalog Number Unit Price Quantity
18 mL 87 mm 24 mm 40954-KT AA40954-KT Each Retrieving
105 mL 59 mm 59 mm 40956-KT AA40956-KT Each Retrieving
190 mL 168 mm 45 mm 40957-KT AA40957-KT Each Retrieving
277 mL 85 mm 73 mm 40958-KT AA40958-KT Each Retrieving
370 mL 133 mm 69 mm 40959-KT AA40959-KT Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Physical Form

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty