Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Low Form Nickel Crucibles, Alfa Aesar

Supplier: Alfa Aesar
35885-KT 35906-KT 35969-KT 36017-KT 35996-KT 36038-KT 35948-KT 36057-KT 35927-KT 36084-KT
In the analytical laboratory, nickel crucibles offer high resistance to dilute alkalies at a very low cost per crucible.

  • Life expectancy of a nickel crucible is from 4 to 6 fusions
  • Low form design
  • Completely resistant to phosphoric acid
  • Highly resistant to the corrosive effect of the strongest alkalies
  • Ideal for use in fusions where zirconium or other metals cannot be used

No dry gases are actively corrosive to nickel at atmospheric temperature. Nickel is also resistant to dry hydrogen chloride, hydrogen fluoride, and chlorine up to about 535°C. Nickel is not affected by steam at temperatures usually encountered. It is corroded by gases containing sulfur.




 

ORDER

Expand All / Collapse All
Capacity Depth Top Diameter Bottom Diameter Supplier No. VWR Catalog Number Unit Price Quantity
20 mL 30 mm 33 mm 25 mm 35906-KT AA35906-KT Each Retrieving
25 mL 23 mm 45 mm 38 mm 35927-KT AA35927-KT Each Retrieving
35 mL 30 mm 46 mm 38 mm 35948-KT AA35948-KT Each Retrieving
45 mL 35 mm 46 mm 38 mm 35969-KT AA35969-KT Each Retrieving
55 mL 43 mm 47 mm 38 mm 35996-KT AA35996-KT Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Nickel Cover for 20 mL Low Form Nickel Crucible 36042-KT AA36042-KT Each Retrieving
Nickel Cover for 25 mL Low Form Nickel Crucible 36082-KT AA36082-KT Each Retrieving
Nickel Cover for 45 mL Low Form Nickel Crucible 36023-KT AA36023-KT Each Retrieving
Nickel Cover for 100 mL Low Form Nickel Crucible 35942-KT AA35942-KT Each Retrieving
Nickel Cover for 250 mL Low Form Nickel Crucible 35921-KT AA35921-KT Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty