Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Low Form Inconel® Alloy 601 Crucibles, Alfa Aesar

Supplier: Alfa Aesar
35907-KT 36056-KT 35886-KT 35994-KT 36016-KT 36037-KT 35949-KT 35928-KT 35970-KT 36085-KT
Inconel® nickel-chromium-iron alloy 601 is a general-purpose engineering material for applications that require resistance to heat and corrosion

  • Ideal for high-temperature applications requiring resistance to oxidation and spalling
  • The alloy is unaffected by rapid changes from hot to cold
  • Retains mechanical strength at elevated temperatures

The high resistance of Inconel® Alloy 601 to oxidation, carburization, or sulfidation make it well suited for vessels used in determining moisture, volatiles, fixed-carbon, and ash located in most coal and coke products, or wood pulp or fiber. It is also recommended for use in drying and ashing biological materials whose residues are soluble in dilute acid or alkali for subsequent analysis. Trace-level determinations of principal constituent elements are excluded. The vessels can be cleaned simply by scouring with sea-sand or some other mild abrasive.

It offers excellent resistance to oxidation in the 1000 to 1200°C temperature range, and provides good corrosion resistance to many acid and aqueous salt solutions.




 

ORDER

Expand All / Collapse All
Capacity Depth Top Diameter Bottom Diameter Supplier No. VWR Catalog Number Unit Price Quantity
15 mL 23 mm 33 mm 25 mm 35886-KT AA35886-KT Each Retrieving
25 mL 23 mm 45 mm 38 mm 35928-KT AA35928-KT Each Retrieving
45 mL 35 mm 46 mm 38 mm 35970-KT AA35970-KT Each Retrieving
55 mL 43 mm 47 mm 38 mm 35994-KT AA35994-KT Each Retrieving
75 mL 43 mm 51 mm 41 mm 36016-KT AA36016-KT Each Retrieving
100 mL 46 mm 59 mm 50 mm 36056-KT AA36056-KT Each Retrieving
250 mL 60 mm 82 mm 66 mm 36085-KT AA36085-KT Each Retrieving
500 mL 66 mm 102 mm 89 mm 36037-KT AA36037-KT Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Iconel® Cover for 15 mL Low Form Iconel® Alloy 601 Crucible 36003-KT AA36003-KT Each Retrieving
Iconel® Cover for 75 mL Low Form Iconel® Alloy 601 Crucible 35962-KT AA35962-KT Each Retrieving
Iconel® Cover for 100 mL Low Form Iconel® Alloy 601 Crucible 35941-KT AA35941-KT Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty