Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Straight Wall Zirconium Crucibles, Alfa Aesar

Supplier: Alfa Aesar
36070-KT 35992-KT 35888-KT 36106-KT 36087-KT 35972-KT 35930-KT 35951-KT 36054-KT 36014-KT 36097-KT 36035-KT 35909-KT
Each zirconium crucible is handmade to an exacting tolerance for uniform wall thickness

  • Special apparatus is not required for handling hot zirconium crucibles
  • Sudden contact with cold, metallic surfaces will have no deleterious effect
  • Zirconium crucibles require a minimum of specialized care; smoothing and shaping is not a special consideration
  • The inherent strength of zirconium precludes the necessity of reinforced rims and thicker bottoms

These crucibles are produced under the most stringent requirements to ensure ultimate tensile strength, yield strength, elongation, and chemical purity.

Only high-purity zirconium material is used.




 

ORDER

Expand All / Collapse All
Capacity Depth O.D. Supplier No. VWR Catalog Number Unit Price Quantity
10 mL 22 mm 27 mm 35909-KT AA35909-KT Each Retrieving
15 mL 22 mm 33 mm 35930-KT AA35930-KT Each Retrieving
20 mL 29 mm 33 mm 35951-KT AA35951-KT Each Retrieving
25 mL 22 mm 45 mm 35972-KT AA35972-KT Each Retrieving
35 mL 29 mm 46 mm 35992-KT AA35992-KT Each Retrieving
45 mL 34 mm 46 mm 36014-KT AA36014-KT Each Retrieving
55 mL 41 mm 47 mm 36070-KT AA36070-KT Each Retrieving
100 mL 45 mm 59 mm 36054-KT AA36054-KT Each Retrieving
250 mL 59 mm 82 mm 36097-KT AA36097-KT Each Retrieving
500 mL 65 mm 101 mm 36087-KT AA36087-KT Each Retrieving
1000 mL 89 mm 127 mm 36035-KT AA36035-KT Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Zirconium Cover for 5 mL Straight Wall Zirconium Crucible 36005-KT AA36005-KT Each Retrieving
Zirconium Cover for 10 mL Straight Wall Zirconium Crucible 36045-KT AA36045-KT Each Retrieving
Zirconium Cover for 15 mL Straight Wall Zirconium Crucible 36079-KT AA36079-KT Each Retrieving
Zirconium Cover for 20 mL Straight Wall Zirconium Crucible 36107-KT AA36107-KT Each Retrieving
Zirconium Cover for 25 mL Straight Wall Zirconium Crucible 36064-KT AA36064-KT Each Retrieving
Zirconium Cover for 45 mL Straight Wall Zirconium Crucible 36111-KT AA36111-KT Each Retrieving
Zirconium Cover for 55 mL Straight Wall Zirconium Crucible 36026-KT AA36026-KT Each Retrieving
Zirconium Cover for 100 mL Straight Wall Zirconium Crucible 35960-KT AA35960-KT Each Retrieving
Zirconium Cover for 250 mL Straight Wall Zirconium Crucible 35939-KT AA35939-KT Each Retrieving
Zirconium Cover for 500 mL Straight Wall Zirconium Crucible 35918-KT AA35918-KT Each Retrieving
Zirconium Cover for 1000 mL Straight Wall Zirconium Crucible 35896-KT AA35896-KT Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty