Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zirconium Crucibles with Snap-On Ring, Alfa Aesar

Supplier: Alfa Aesar
35966-KT 36020-KT 35903-KT
Zirconium's distinctive properties make it a popular choice for fusions and fluxes in analytical chemistry and chemical processing.

  • Crucibles feature a snap-on ring
  • Removable ring snap-fits into a machined groove approximately 0.20" below the top of the crucible

Fluxes that can be used in these zirconium crucibles include Sodium Peroxide Fusion, Sodium Carbonate Fusion, and Lithium Salt Fusion. Prolonged exposure to air at temperatures of more than 750°C can have a negative effect on zirconium.




 

ORDER

Expand All / Collapse All
Height Top Diameter Bottom Diameter O.D. Supplier No. VWR Catalog Number Unit Price Quantity
33 mm 39 mm 29 mm 44 mm Ring/Rim 35903-KT AA35903-KT Each Retrieving
44 mm 42 mm 29 mm 48 mm Ring/Rim 36020-KT AA36020-KT Each Retrieving
36 mm 47 mm 38 mm 50 mm Ring/Rim 35966-KT AA35966-KT Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty