TUNE SpectraMax® Detection Cartridge, Molecular Devices
0200-7050POS 0200-7050POS
10830-228EA 34459.48 USD
10830-228
TUNE SpectraMax® Detection Cartridge, Molecular Devices
The TUNE technology combines the wavelength flexibility of a monochromator-based system with the signal strength of a filter design.
  • Wavelength flexibility
  • Strong signal
  • High-powered light source
  • TUNE technology
High-powered LEDs provide an adjustable light source, while the TUNE technology selects the excitation and emission wavelengths of choice.
TUNE SpectraMax® Detection Cartridges for the SpectraMax® Paradigm® System offer unmatched sensitivity in wavelength tunable detection. Designed to work with top- or bottom-read, it supports detection in Fluorescence Intensity (FI), Time-Resolved Fluorescence (TRF), and Luminescence (LUM).
The optimization wizard in SoftMax® Pro Software quickly scans excitation and emission spectra to select the ideal settings for the user’s assay.