Human Recombinant HCC1 (from E. coli)
Supplier: BioVision
4110-1000 4110-10
10005-866EA 4494.85 USD
10005-866 10005-864
Human Recombinant HCC1 (from E. coli)
A cytokine and a possible splicing factor
The biological activity of HCC-1 was determined by a chemotaxis assay using human monocytes. The maximum activity was achieved at 500 ng/ml.
Human HerpatoCellular Carcinoma (HCC-1) was first isolated from the hemofiltrate of patients with chronic renal failure. HCC-1 has a 46% amino acid sequence identity with MIP-1 α/MIP-1 β and 29%-37% homology with other CC chemokines. The human HCC-1 is an 8.4 kDa protein containing 72 amino acid residues.