Human Recombinant FGF9 (from E. coli), BioVision
Supplier: BioVision
4056-1000 4056-20
10005-774EA 221.82 USD
10005-774 10005-772
Human Recombinant FGF9 (from E. coli), BioVision
A heparin binding growth factor, stimulating the proliferation and activation of cells expressing FGFR
The ED₅₀ as determined by the dose-dependent stimulation of thymidine uptake by BaF3 cells expressing FGF receptors is <0.5 ng/ml, corresponding to a specific activity of >2 x 10⁶ units/mg.
FGF-9 (also known as Glial Activating Factor) is a heparin binding growth factor that stimulates the proliferation and activation of glial cells and other cells that express FGF receptors. The human FGF-9 is a 23.4 kDa protein containing 207 amino acid residues.