PF-573228

Supplier: BioVision

Synonyms: 6-((4-((3-(Methylsulfonyl)benzyl)amino)-5-(trifluoromethyl)pyrimidin-2-yl)amino)-3,4-dihydroquinolin-2(1H)-one6-((4-((3-(Methylsulphonyl)benzyl)amino)-5-(trifluoromethyl)pyrimidin-2-yl)amino)-3,4-dihydroquinolin-2(1H)-one

AB273523-50MG AB273523-10MG
10003-140EA 397.75 USD
10003-140 10003-142
PF-573228
PF-573228
Formula: C₂₂H₂₀F₃N₅O₃S
MW: 491.4861 g/mol
MDL Number: MFCD11519967
CAS Number: 869288-64-2

Order Now

Specification Test Results

Purity ≥98% by HPLC
Form Off-white solid
CAS 869288-64-2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR