WST-3

Supplier: BioVision

AB285431-100MG AB285431-500MG AB285431-1G
10003-124EA 337.08 USD
10003-124 10003-126 10003-128
WST-3
WST-3
Formula: C₁₉H₁₀IN₆NaO₁₀S₂
CAS Number: 515111-36-1

Order Now

Specification Test Results

Purity ≥98% by HPLC
Form Pale yellow to pale brown solid
CAS 515111-36-1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR