Nanaomycin A ≥98% (by HPLC)

2790-1
75836-806EA 159.53 USD
75836-806
Nanaomycin A ≥98% (by HPLC)
Nanaomycin A

Nanaomycin A is a selective DNA methyltransferase 3B (DNMT3B) inhibitor


  • Crystalline Solid


It induces antiproliferative effects in three different tumor cell lines originating from different tissues. Nanaomycin A treatment reduces the global methylation levels in all three cell lines and reactivates transcription of the RASSF1A tumor suppressor gene.


DMSO (~10 mg/mL)

Storage Temperature: Freezer
CAS Number: 52934-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR