AS601245
Synonyms:
2-(Benzo[d]thiazol-2-yl)-2-(2-((2-(pyridin-3-yl)ethyl)amino)pyrimidin-4-yl)acetonitrile
2162-5
2162-1
10191-492EA
286.04
USD
10191-492
10191-490
AS601245
AS601245
Formula:
C₂₀H₁₆N₆S MW: 372,45 g/mol |
MDL Number:
MFCD07772196 CAS Number: 345987-15-7 |
Specification Test Results
Purity | ≥95% by HPLC |
Form | White to yellow solid |
CAS | 345987-15-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...