Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Grand Refrigerators, Glass Door, Nor-Lake®

Supplier: NOR-LAKE SCIENTIFIC
GR49SSG/0 GR23SSG/0 GR72SSG/0
Nor-Lake® Grand Refrigerator [+4°C (+39°F)]

SS Int, SS Ext, Digital LED display microprocessor temperature controller, Selectable °F or °C control, Visual Hi/Lo temperature alarm, Adjustable control range from +2°C to +10°C (+35.6°F to +50°F) ; factory preset at +4°C (+39°F), Door open alarm, Continuous temperature display, Condenser cleaning reminder alarm, Power supply switch, Exterior cabinet front and sides are stainless steel, Exterior cabinet top, back and bottom are galvanized steel, Interior cabinet liner is stainless steel, Cabinet is foamed-in-place with CFC-free high density polyurethane foam insulation, Full-length glass door(s) with pivot hinges Self-closing with 90° stay open feature, Magnetic door gaskets, Top mounted key door lock(s) (2 keys), 4 casters (2 locking) on one and two door models, 6 casters (3 locking) on three door model, Field installation of casters required, 4 vinyl coated wire shelves per door section, Perimeter anti-condensate door frame heater wires, Interior light, switch activated, Bottom mounted air cooled condensing unit, Top mounted forced air cooling coil, Automatic defrost and condensate evaporation, Warranties: 18 month parts and labor, 5 year, compressor (US and Canada), 18 months parts

Ordering information: Accessories not included. All Units include 18 months limited warranty on parts and labor ( US & Canada)




 

ORDER

Expand All / Collapse All
Volume Dimensions No. of Doors Supplier No. VWR Catalog Number Unit Price Quantity
650 L (23 cu. ft.) 27 1/2 W x 83 3/4 H x 33 D" 1 GR23SSG/0 10803-776 Each Retrieving
UL Listed cUL Listed
1388 L (49 cu. ft.) 55 1/4 W x 83 3/4 H x 33D" 2 GR49SSG/0 10803-778 Each Retrieving
UL Listed cUL Listed
1982 L (70 cu. ft.) 78W x 83 3/4 H x 33D" 3 GR72SSG/0 10803-734 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Electrical 115V, 5-15P
Temperature Range 2 to 10°C (35.6 to 50°F)

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty