Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Stability Chamber-Refrigerated Incubators, Glass Door, Nor-Lake® Scientific

Supplier: NOR-LAKE SCIENTIFIC
NSRI241WSG/4 NSRI522WSG/5 NSRI331WSG/4 NSRI241WSG/5 NSRI331WSG/5 NSRI241WSG/0 NSRI803WSG/4 NSRI803WSG/5 NSRI803WSG/0 NSRI522WSG/0 NSRI522WSG/4 NSRI331WSG/0
Nor-Lake® Scientific Stability Chamber-Refrigerated Incubator

Exterior front, sides and back powder coated. Exterior top and bottom glavanized steel. Interior cabinet is stainless steel.

Digital 132 x 64 LED display, Programmable logic (PLC) microprocessor temperature controller, Audible and visual Hi/Lo temperature alarm, Adjustable control range from +20°C to +60°C, Programmable ramp and soak, Remote alarm contacts, Continuous product temperature display, 3 sensor system; 1 product, 1 air, 1 coil, Sensor access port 1" diameter, Power supply breaker and convenience power switch located on facade, Door ajar alarm, audible and visual, Temperature transmitter (0-10 VDC) (selectable to air or product), RS-485 digital communications port (MODBUS-RTU), Password protection - set-points and factory settings, 100 event alarm logging date/time stamp, Integrated Hi/Lo product temperature alarm test, Digital calibration air and product, Real time clock with date and time display, Door opening counter with 24 hour automatic reset, Adjustable alarm beep or constant on (key pad selectable), Low battery test switch with buzzer, Perimeter anti-condensate door frame heater (on/off temperature selectable), Sensor failure alarm, Power failure alarm, Exterior cabinet front, sides and back are powder coated, Exterior cabinet top and bottom are galvanized steel, Interior cabinet liner is stainless steel, Cabinet is foamed-in-place with CFC-free high density, polyurethane foam insulation, Glass door(s), full size, Heavy duty door pivot hinges, Pull door handles, full length, Magnetic door gaskets, Centered key door lock(s) (2 keys), Four casters (2 locking) on one and two door models, six casters (3 locking) on three door models, 3 coated wire shelves per door section, Air cooled condensing unit, Top mounted forced air refrigeration system, Condensate evaporation, LED lighting, Warranties: 18 months parts and labor, 5 year compressor, (US and Canada), 18 months parts .

Ordering information: Accessories not included. All Units include 18 months limited warranty on parts and labor (US & Canada).




 

ORDER

Expand All / Collapse All
Description Electrical Volume Dimensions Temperature Range Supplier No. VWR Catalog Number Unit Price Quantity
Incubator with One Glass Door 115V, 5-20P 24 cu. ft. 27 1/2 W x 79 5/8 H x 34 7/8 D" 20 to 60°C NSRI241WSG/0 10768-454 Each Retrieving
UL Listed cUL Listed
Incubator with One Glass Door 208-230V, 6-15P 24 cu. ft. 27 1/2 W x 79 5/8 H x 34 7/8 D" 20 to 60°C NSRI241WSG/4 10769-068 Each Retrieving
UL Listed cUL Listed
Incubator with One Glass Door 230V, 1P, 50HZ Power Inlet (IEC 60320) Module 24 cu. ft. 27 1/2 W x 79 5/8 H x 34 7/8 D" 20 to 60°C NSRI241WSG/5 10769-070 Each Retrieving
Incubator with One Glass Door 115V, 5-20P 33 cu. ft. 31 3/4 W x 87 5/8 H x 35 7/8 D" 20 to 60°C NSRI331WSG/0 10768-640 Each Retrieving
UL Listed cUL Listed
Incubator with One Glass Door 208-230V, 6-15P 33 cu. ft. 31 3/4 W x 87 5/8 H x 35 7/8 D" 20 to 60°C NSRI331WSG/4 10768-450 Each Retrieving
UL Listed cUL Listed
Incubator with One Glass Door 230V, 1P, 50HZ Power Inlet (IEC 60320) Module 33 cu. ft. 31 3/4 W x 87 5/8 H x 35 7/8 D" 20 to 60°C NSRI331WSG/5 10768-452 Each Retrieving
Incubator with Two Glass Doors 208-230V, 6-15P 52 cu. ft. 55 W x 79 5/8 H x 34 7/8 D" 20 to 60°C NSRI522WSG/4 10768-480 Each Retrieving
UL Listed cUL Listed
Incubator with Two Glass Doors 230V, 1P, 50HZ Power Inlet (IEC 60320) Module 52 cu. ft. 55 W x 79 5/8 H x 34 7/8 D" 20 to 60°C NSRI522WSG/5 10768-482 Each Retrieving
Incubator with Three Glass Doors 115V, 5-20P 80 cu. ft. 82 1/2 W x 79 5/8 H x 34 7/8 D" 20 to 60°C NSRI803WSG/0 10769-072 Each Retrieving
UL Listed cUL Listed
Incubator with Three Glass Doors 208-230V, 6-15P 80 cu. ft. 82 1/2 W x 79 5/8 H x 34 7/8 D" 20 to 60°C NSRI803WSG/4 10768-456 Each Retrieving
UL Listed cUL Listed
Incubator with Three Glass Doors 230V, 1P, 50HZ Power Inlet (IEC 60320) Module 80 cu. ft. 82 1/2 W x 79 5/8 H x 34 7/8 D" 20 to 60°C NSRI803WSG/5 10768-458 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty