Human Recombinant IL7 (from E. coli)
Supplier: BioVision
4146-50
10006-180EA 982.75 USD
10006-180
Human Recombinant IL7 (from E. coli)
Proteins and Peptides
A cytokine important for B and T cell development.
The ED₅₀ as determined by the dose-dependent stimulation of the proliferation of murine 2E8 cells is <0.5 ng/ml, corresponding to a specific activity of >2 x 10⁶ units/mg.
IL-7 is a hematopoietic growth factor which affects primarily early B and T cells. Produced by thymic stromal cells, spleen cells and keratinocytes, IL-7 can also co-stimulate the proliferation of mature T cells in combination with other factors such as ConA and IL-2. Human and murine IL-7 is cross-species reactive. Recombinant human IL-7 is a 17.4 kDa protein containing 153 amino acid residues.