Human Recombinant IL5 (from E. coli)
Supplier: BioVision
4140-1000 4140-10
10006-154EA 5779.09 USD
10006-154 10006-152
Human Recombinant IL5 (from E. coli)
A cytokine which stimulates B cell growth & eosinophil activation.
The ED₅₀ as determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is <0.15 ng/ml, corresponding to a specific activity of >6 x 10⁶units/mg.
IL-5 is a potent T-cell derived factor that stimulates the growth of certain B cells and the differentiation of eosinophils. Human IL-5 is a 26.5 kDa protein containing 232 amino acid residues.