The VWR® Plus Series Glass Door Laboratory Refrigerators are purpose built for scientific and medical applications.
- Adjustable operating temperature range: 1 to 10°C (33.8 to 50°F)
- Sample simulated digital temperature display
- Audible and visual high/low temperature alarms
- Swivel casters add four inches to height
Units feature a digital microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies.
The all new temperature display and alarm module offers industry leading product security with a monitoring and alarm system that is 100% independent of the temperature controller. Real time sample simulated temperatures are constantly displayed and supported by audible/visual high and low temperature alarms, along with power outage and sensor error alarms. Remote alarm dry contacts and an access port are provided to simplify connections to existing facility monitoring systems.
Environmentally friendly, natural hydrocarbon (HC) refrigerants (not containing hydrofluorocarbons) vastly reduce global-warming potential (GWP) while meeting new EPA/SNAP mandates, as well as UL, ASHRAE compliance guidelines.
A forced air directional refrigeration system provides superior temperature uniformity and recovery after door openings.
Certifications: UL/C-UL listed.
Ordering information: Two-year parts and labor warranty, five years on compressor parts.