Anti-IL3RA Rat Monoclonal Antibody [clone: 151231]

Supplier: R & D Systems
MAB983
220021-740EA 772.25 USD
220021-740
Anti-IL3RA Rat Monoclonal Antibody [clone: 151231]
Antibodies
Type: Primary
Antigen: IL3RA
Clonality: Monoclonal
Clone: 151231
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR