Anti-CD14 Rat Monoclonal Antibody [clone: 159010]

Supplier: R & D Systems
MAB982
220021-739EA 763.57 USD
220021-739
Anti-CD14 Rat Monoclonal Antibody [clone: 159010]
Antibodies
Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 159010
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR