Anti-CXCL16 Rat Monoclonal Antibody [clone: 256213]

Supplier: R & D Systems
MAB976
220021-735EA 600.15 USD
220021-735
Anti-CXCL16 Rat Monoclonal Antibody [clone: 256213]
Antibodies
Type: Primary
Antigen: CXCL16
Clonality: Monoclonal
Clone: 256213
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR