Anti-IL1RAPL1 Mouse Monoclonal Antibody [clone: 143116]

Supplier: R & D Systems
MAB975
220021-734EA 763.57 USD
220021-734
Anti-IL1RAPL1 Mouse Monoclonal Antibody [clone: 143116]
Antibodies
Type: Primary
Antigen: IL1RAPL1
Clonality: Monoclonal
Clone: 143116
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR