Anti-TIMP4 Mouse Monoclonal Antibody [clone: 153934]

Supplier: R & D Systems
MAB974
220021-733EA 772.25 USD
220021-733
Anti-TIMP4 Mouse Monoclonal Antibody [clone: 153934]
Antibodies
Type: Primary
Antigen: TIMP4
Clonality: Monoclonal
Clone: 153934
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR